Anti-CDKN1C Rabbit Polyclonal Antibody

Supplier: Abnova
PAB18370
10707-384EA 426.15 USD
10707-384
Anti-CDKN1C Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CDKN1C.

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunofluorescence: 1:500-1:1000; ELISA: 1:5000

Type: Primary
Antigen: CDKN1C
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR