Anti-PDGFRB Rabbit Polyclonal Antibody

Supplier: Abnova
PAB18373
10728-738EA 426.15 USD
10728-738
Anti-PDGFRB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of PDGFRB.

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:100; Immunofluorescence: 1:500-1:1000; ELISA: 1:5000

Type: Primary
Antigen: PDGFRB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR