Anti-F11R Goat Polyclonal Antibody

Supplier: Abnova
PAB18709
10712-794EA 527.69 USD
10712-794
Anti-F11R Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of F11R.

Recommended Dilutions: ELISA: 1:2000; Western Blot: 0.3-2 ?g/ml

Type: Primary
Antigen: F11R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR