Anti-MHC2 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: M5/114]

Supplier: Abnova
MAB9819
10725-462EA 426.15 USD
10725-462
Anti-MHC2 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: M5/114]
Antibodies
Rat monoclonal antibody raised against MHC Class II.

Recommended Dilutions: Flow Cytometry: 4 ?g/ml

Type: Primary
Antigen: MHC2
Clonality: Monoclonal
Clone: M5/114
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR