Anti-STC2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB3090
10680-106EA 406.15 USD
10680-106
Anti-STC2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of STC2.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500 Immunohistochemistry: 1:50-100

Type: Primary
Antigen: STC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR