Anti-CDH13 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB3081
10678-414EA 406.15 USD
10678-414
Anti-CDH13 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CDH13.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:50-100; Immunohistochemistry: 1:10-50; Flow Cytometry: 1:10-50

Type: Primary
Antigen: CDH13
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR