Anti-IL2RB Rabbit Polyclonal Antibody

Supplier: Abnova
PAB18399
10719-228EA 426.15 USD
10719-228
Anti-IL2RB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of IL2RB.

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: IL2RB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR