Anti-ITPR1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB18402
10719-908EA 426.15 USD
10719-908
Anti-ITPR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ITPR1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Immunofluorescence: 1:500-1:1000; ELISA: 1:20000

Type: Primary
Antigen: ITPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR