Anti-IQGAP1 Goat Polyclonal Antibody

Supplier: Abnova
PAB18725
10719-492EA 527.69 USD
10719-492
Anti-IQGAP1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of IQGAP1.

Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: IQGAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR