Anti-IKZF1 Goat Polyclonal Antibody

Supplier: Abnova
PAB18736
10718-992EA 527.69 USD
10718-992
Anti-IKZF1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of IKZF1.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1-3 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2.5-3.8 ?g/ml

Type: Primary
Antigen: IKZF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR