Anti-UBP1 Goat Polyclonal Antibody

Supplier: Abnova
PAB18748
10743-692EA 527.69 USD
10743-692
Anti-UBP1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of UBP1.

Recommended Dilutions: ELISA: 1:2000

Type: Primary
Antigen: UBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR