Anti-GATA6 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB4926
10680-574EA 426.15 USD
10680-574
Anti-GATA6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GATA6.

Recommended Dilutions: ELISA: 1:1000; Dot Blot: 1:500

Type: Primary
Antigen: GATA6
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR