Anti-CDH1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 67A4]

Supplier: Abnova
MAB9850
10707-208EA 406.15 USD
10707-208
Anti-CDH1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 67A4]
Antibodies
Mouse monoclonal antibody raised against CDH1.

Recommended Dilutions: Flow Cytometry: 10 ?l reagent/100 ?l of whole blood or 10^6 cells

Type: Primary
Antigen: CDH1
Clonality: Monoclonal
Clone: 67A4
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR