Anti-SLC39A7 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB19352
10738-604EA 527.69 USD
10738-604
Anti-SLC39A7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody rasied against synthetic peptide of SLC39A7.

Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; Immunohistochemistry: 2.5 ?g/ml

Type: Primary
Antigen: SLC39A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR