Anti-ITGAM Rat Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: 3A33]

Supplier: Abnova
MAB9860
10719-720EA 426.15 USD
10719-720
Anti-ITGAM Rat Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: 3A33]
Antibodies
Rat monoclonal antibody raised against Itgam.

Recommended Dilutions: Flow Cytometry: <0.3 ?g/10^6 cells

Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: 3A33
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR