Anti-ZNF281 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB19360
10745-546EA 527.69 USD
10745-546
Anti-ZNF281 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ZNF281.

Recommended Dilutions: Western Blot: 0.25-0.5 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: ZNF281
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR