Anti-ITGAM Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7® ) [clone: M1/70]

Supplier: Abnova
MAB9867
10719-756EA 406.15 USD
10719-756
Anti-ITGAM Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7® ) [clone: M1/70]
Antibodies
Rat monoclonal antibody raised against Itgam.

Recommended Dilutions: Flow Cytometry: <0.3 ?g/10^6 cells

Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: M1/70
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR