Anti-4-pregnen-17, 20-diol-3-one Polyclonal Antibody

Supplier: Abnova
PAB5171
10672-582EA 406.15 USD
10672-582
Anti-4-pregnen-17, 20-diol-3-one Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against 4-pregnen-17, 20-diol-3-one.

Type: Primary
Antigen: 4-pregnen-17, 20-diol-3-one
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR