Anti-CSF1R Goat Polyclonal Antibody

Supplier: Abnova
PAB18945
10709-210EA 527.69 USD
10709-210
Anti-CSF1R Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of CSF1R.

Recommended Dilutions: ELISA: 1:8000; Western Blot: 0.2-1 ?g/ml

Type: Primary
Antigen: CSF1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR