Anti-ACVR1B Rabbit Polyclonal Antibody

Supplier: Abnova
PAB18053
10688-056EA 426.15 USD
10688-056
Anti-ACVR1B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ACVR1B.

  • Recommended Dilutions: Western Blot: 1:500~1:1000; Immunofluorescence: 1:500~1:1000; ELISA: 1:5000

Type: Primary
Antigen: ACVR1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR