Anti-GJB2 Goat Polyclonal Antibody

Supplier: Abnova
PAB7162
10720-574EA 527.69 USD
10720-574
Anti-GJB2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of GJB2.

Recommended Dilutions: ELISA: 1:128000; Western Blot: 0.01-0.03 ?g/ml

Type: Primary
Antigen: GJB2
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Goat
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR