Anti-Quinolones (Fluoroquinolones) Polyclonal Antibody

PAB8328
10730-888EA 647.69 USD
10730-888
Anti-Quinolones (Fluoroquinolones) Polyclonal Antibody
Sheep polyclonal antibody raised against Quinolones.

Recommended Dilutions: Enzyme Immunoassay (5 ?g/ml

Type: Primary
Antigen: Quinolones (Fluoroquinolones)
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR