Anti-VPS25 Goat Polyclonal Antibody

Supplier: Abnova
PAB6937
10744-276EA 527.69 USD
10744-276
Anti-VPS25 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of VPS25.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 0.5-1.5 ?g/ml; Immunohistochemistry: 10 ?g/ml

Type: Primary
Antigen: VPS25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR