Anti-AKT1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB7912
10674-200EA 527.69 USD
10674-200
Anti-AKT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of AKT1.

Recommended Dilutions: ELISA: 1:2000; Western Blot: 1:500

Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR