Anti-UBE2V1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9746
10743-648EA 920 USD
10743-648
Anti-UBE2V1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of UBE2V1.

Type: Primary
Antigen: UBE2V1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR