Anti-DAPK3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9749
10710-202EA 920 USD
10710-202
Anti-DAPK3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of DAPK3.

Type: Primary
Antigen: DAPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR