Anti-SMPD1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9751
10738-744EA 786.15 USD
10738-744
Anti-SMPD1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of SMPD1.

Type: Primary
Antigen: SMPD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR