Anti-BACE2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9773
10690-692EA 920 USD
10690-692
Anti-BACE2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of BACE2.

Type: Primary
Antigen: BACE2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR