Anti-SH3BP2 Sheep Polyclonal Antibody

Supplier: Abnova
PAB9781
10677-940EA 527.69 USD
10677-940
Anti-SH3BP2 Sheep Polyclonal Antibody
Antibodies
Sheep polyclonal antibody raised against partial recombinant Sh3bp2.

Type: Primary
Antigen: SH3BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR