Anti-RNF216 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9789
10731-790EA 920 USD
10731-790
Anti-RNF216 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Rnf216.

Type: Primary
Antigen: RNF216
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR