Anti-BrdU 0 Polyclonal Antibody

Supplier: Abnova
PAB9791
10682-038EA 653.85 USD
10682-038
Anti-BrdU 0 Polyclonal Antibody
Antibodies
Sheep polyclonal antibody raised against BrdU.

Recommended Dilutions: Western Blot: 25-100 ?g/ml; Immunoprecipitation: 25-100 ?g/ml; Immunohistochemistry: 25-100 ?g/ml

Type: Primary
Antigen: BrdU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: 0
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR