Anti-CACNA1A Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9793
10704-624EA 527.69 USD
10704-624
Anti-CACNA1A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Cacna1a.

Type: Primary
Antigen: CACNA1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR