Anti-CASP9 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9830
10705-266EA 920 USD
10705-266
Anti-CASP9 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CASP9.

Recommended Dilutions: Western Blot: 0.25-0.5 ?g/lml

Type: Primary
Antigen: CASP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR