Anti-CHEK1 Sheep Polyclonal Antibody

Supplier: Abnova
PAB9841
10677-830EA 527.69 USD
10677-830
Anti-CHEK1 Sheep Polyclonal Antibody
Antibodies
Sheep polyclonal antibody raised against partial recombinant CHEK1.

Recommended Dilutions: Western Blot: 1-10 ?g/ml

Type: Primary
Antigen: CHEK1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR