Anti-CX3CR1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9848
10709-542EA 920 USD
10709-542
Anti-CX3CR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CX3CR1.

Recommended Dilutions: Western Blot: 0.5-1.0 ?g/ml

Type: Primary
Antigen: CX3CR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR