Anti-EIA Sheep Polyclonal Antibody

Supplier: Abnova
PAB9863
10711-762EA 527.69 USD
10711-762
Anti-EIA Sheep Polyclonal Antibody
Antibodies
Sheep polyclonal antibody raised against a recombinant EIA.

Recommended Dilutions: Western Blot: 1-10 ?g/ml

Type: Primary
Antigen: EIA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR