Anti-EPHB1 Sheep Polyclonal Antibody

Supplier: Abnova
PAB9865
10677-850EA 527.69 USD
10677-850
Anti-EPHB1 Sheep Polyclonal Antibody
Antibodies
Sheep polyclonal antibody raised against partial recombinant EPHB1.

Recommended Dilutions: Western Blot: 1-10 ?g/ml; Immunoprecipitation: 10-20 ?g/ml

Type: Primary
Antigen: EPHB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR