Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ALOX5AP Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9875
10688-844
Anti-ALOX5AP Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of ALOX5AP.

Recommended Dilutions: Western Blot: 1-5 μg/ml

Type: Primary
Antigen: ALOX5AP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty