Anti-CACNG2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9884
10705-414EA 527.69 USD
10705-414
Anti-CACNG2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Cacng2.

Recommended Dilutions: Western Blot: 5-10 ?g/ml

Type: Primary
Antigen: CACNG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR