Anti-GFRA2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9890
10715-834EA 920 USD
10715-834
Anti-GFRA2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of GFRA2.

Recommended Dilutions: Western Blot: 0.25-0.5 ?g/lml

Type: Primary
Antigen: GFRA2
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR