Anti-GNAI1/GNAI2 Polyclonal Antibody

Supplier: Abnova
PAB9893
10716-028EA 527.69 USD
10716-028
Anti-GNAI1/GNAI2 Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of GNAI1/GNAI2.

Recommended Dilutions: Western Blot: 5-10 ?g/ml

Type: Primary
Antigen: GNAI1/GNAI2
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR