Anti-P2RY13 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9895
10728-300EA 527.69 USD
10728-300
Anti-P2RY13 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of P2RY13.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: P2RY13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR