Anti-Farnesyl Polyclonal Antibody

Supplier: Abnova
PAB9904
10714-384EA 653.85 USD
10714-384
Anti-Farnesyl Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against Farnesyl.

Type: Primary
Antigen: Farnesyl
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR