Anti-EGO1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9915
10711-730EA 786.15 USD
10711-730
Anti-EGO1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ego-1.

Recommended Dilutions: ELISA: 1:25000-1:100000; Western Blot: 1:500-1:5000

Type: Primary
Antigen: EGO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Nematoda
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR