Anti-ATR Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9924
10690-520EA 786.15 USD
10690-520
Anti-ATR Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ATR.

Recommended Dilutions: ELISA: 1:15000-1:70000; Western Blot: 1:1000-1:5000

Type: Primary
Antigen: ATR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Clawed frog, Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR