Anti-ABCB5 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9925
10687-618EA 786.15 USD
10687-618
Anti-ABCB5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ABCB5.

Recommended Dilutions: ELISA: 1:80000-1:85000; Western Blot: 1:10000

Type: Primary
Antigen: ABCB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR