Anti-ABCB6 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9926
10687-622EA 786.15 USD
10687-622
Anti-ABCB6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ABCB6.

Recommended Dilutions: ELISA: 1:40000-1:200000; Western Blot: 1:1000-1:5000

Type: Primary
Antigen: ABCB6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR