Anti-AHSA1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9927
10688-562EA 786.15 USD
10688-562
Anti-AHSA1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of AHSA1.

Recommended Dilutions: ELISA: 1:35000-1:185000; Western Blot: 1:500-1:3000

Type: Primary
Antigen: AHSA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR