Anti-AGAP2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9929
10688-410EA 786.15 USD
10688-410
Anti-AGAP2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against partial recombinant AGAP2.

  • Recommended Dilutions: ELISA: 1:40000-1:200000; Western Blot: 1:5000-1:20000

Type: Primary
Antigen: AGAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR