Anti-AKT2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB9933
10692-404EA 786.15 USD
10692-404
Anti-AKT2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of AKT2.

Recommended Dilutions: ELISA: 1:4000-1:16000; Western Blot: 1:1000

Type: Primary
Antigen: AKT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR